Skip to Content
Merck

Skip To

34533

Fensulfothion oxon

PESTANAL®, analytical standard

Synonym(s):

Diethyl 4-(methylsulfinyl)phenyl phosphate, Fensulfothion PO-sulfoxide, NSC 133045

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
CZK 8,680.00

About This Item

Empirical Formula (Hill Notation):
C11H17O5PS
CAS Number:
Molecular Weight:
292.29
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
77101502
MDL number:

CZK 8,680.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CS(C1=CC=C(OP(OCC)(OCC)=O)C=C1)=O

InChI

1S/C11H17O5PS/c1-4-14-17(12,15-5-2)16-10-6-8-11(9-7-10)18(3)13/h6-9H,4-5H2,1-3H3

InChI key

GNTVZNNILZKEIB-UHFFFAOYSA-N

Application

Fensulfothion oxon may be used as a reference standard in the determination of fensulfothion oxon in baby food using high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS/MS) and ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS).

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
892573237332374
format

neat

format

neat

format

neat

format

neat

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

product line

PESTANAL®

product line

PESTANAL®

product line

PESTANAL®

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

application(s)

agriculture
environmental

application(s)

-

application(s)

agriculture
environmental

application(s)

agriculture
environmental


pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters