Skip to Content
Merck

Skip To

34161

Sudan IV-d6

analytical standard

Synonym(s):

1-[2-Methyl-4-(2-methyl-phenylazo)phenylazo]-2-naphthol-d6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
CZK 13,100.00

About This Item

Empirical Formula (Hill Notation):
C24D6H14N4O
CAS Number:
Molecular Weight:
386.48
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:

CZK 13,100.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c2c(\N=N\c3ccc(cc3C)\N=N\c4ccccc4C)c(O)c([2H])c([2H])c2c1[2H]

InChI

1S/C24H20N4O/c1-16-7-3-6-10-21(16)26-25-19-12-13-22(17(2)15-19)27-28-24-20-9-5-4-8-18(20)11-14-23(24)29/h3-15,29H,1-2H3/b26-25+,28-27+/i4D,5D,8D,9D,11D,14D

InChI key

RCTGMCJBQGBLKT-GDXALTGDSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sudan IV-d6 may be used as an internal standard for the determination of azo dyes in hot chilli food samples by pressurised liquid extraction and liquid chromatography with electrospray ionization-tandem mass spectrometry.(4)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
341845138368562
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

format

neat

format

neat

format

neat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

mass shift

M+6

mass shift

-

mass shift

-

mass shift

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters