Skip to Content
Merck

Skip To

33869

2-NP-AMOZ

VETRANAL®, analytical standard

Synonym(s):

5-(Morpholinomethyl)-3-(2-nitrobenzylidenamino)-2-oxazolidinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
CZK 4,400.00

About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 4,400.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[O-][N+](=O)c1ccccc1\C=N\N2CC(CN3CCOCC3)OC2=O

InChI

1S/C15H18N4O5/c20-15-18(11-13(24-15)10-17-5-7-23-8-6-17)16-9-12-3-1-2-4-14(12)19(21)22/h1-4,9,13H,5-8,10-11H2/b16-9+

InChI key

PCYAMVONOKWOLP-CXUHLZMHSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
2-NP-AMOZ may be used as a reference standard in the determination of 2-NP-AMOZ in aquatic products using ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS).

Other Notes

Derivatized standard for the assay of metabolites of furazolidone and furaltadone in animal tissue by LC/MS[1]

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
333493386833881
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

format

neat

format

neat

format

neat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters