Skip to Content
Merck
All Photos(1)

Documents

32831

Supelco

2,4-Diaminoanisole

analytical standard

Synonym(s):

4-Methoxy-m-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NH2)2
CAS Number:
Molecular Weight:
138.17
Colour Index Number:
76050
Beilstein:
2085522
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(N)cc1N

InChI

1S/C7H10N2O/c1-10-7-3-2-5(8)4-6(7)9/h2-4H,8-9H2,1H3

InChI key

BAHPQISAXRFLCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Diaminoanisole belongs to the class of primary aromatic amines (PAAs). It is categorized as a carcinogenic compound.

Application

2,4-Diaminoanisole may be used as an analytical reference standard for the determination of the analyte in:
  • Mainstream waterpipe smoke using liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS).
  • Haircolor products by high performance liquid chromatography (HPLC).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Simultaneous quantitative determination of resorcinol and 1-naphthol in haircolor products by high-performance liquid chromatography.
Wang LH and Kuo YP
Chromatographia, 49(3-4) (1999)
Analysis of primary aromatic amines in the mainstream waterpipe smoke using liquid chromatography-electrospray ionization tandem mass spectrometry.
Schubert J
Journal of Chromatography A, 1218(33), 5628-5637 (2011)
Christopher I Olivares et al.
Journal of hazardous materials, 304, 214-221 (2015-11-10)
Recent studies have begun to assess the environmental fate and toxicity of 2,4-dinitroanisole (DNAN), an insensitive munition compound of interest to defense agencies. Aerobic and anaerobic DNAN biotransformation in soils was evaluated in this study. Under aerobic conditions, there was
Emese Hadnagy et al.
Environmental science and pollution research international, 25(24), 24403-24416 (2018-06-18)
The manufacturing of insensitive munition 2,4-dinitroanisole (DNAN) generates waste streams that require treatment. DNAN has been treated previously with zero-valent iron (ZVI) and Fe-based bimetals. Use of Mg-based bimetals offers certain advantages including potential higher reactivity and relative insensitivity to
Matthew J Berens et al.
Environmental science. Processes & impacts, 21(1), 51-62 (2018-11-30)
The recent development of insensitive munitions, such as 2,4-dinitroanisole (DNAN), as components of military explosives has generated concern for potential subsurface contamination and created a need to fully characterize their transformation processes. Compound specific isotope analysis (CSIA) has proven to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service