Skip to Content
Merck
All Photos(1)

Documents

32758

Supelco

Zearalenone-13C18 solution

~25 μg/mL in acetonitrile, analytical standard

Synonym(s):

Fully 13C-labelled Zearalenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C18H22O5
CAS Number:
Molecular Weight:
336.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~25 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+18

storage temp.

−20°C

SMILES string

[13CH3][13C@H]1[13CH2][13CH2][13CH2][13C](=O)[13CH2][13CH2][13CH2]\[13CH]=[13CH]\[13c]2[13cH][13c](O)[13cH][13c](O)[13c]2[13C](=O)O1

InChI

1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/b7-3+/t12-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1

InChI key

MBMQEIFVQACCCH-NMEASMPJSA-N

General description

Certan Vial
Zearalenone-13C18 is the 13C isotope-labeled analog of the mycotoxin, zearalenone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Zearalenone-13C18 may be used as an analytical standard for the determination of zearalenone in animal feed by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).

Analysis Note

purity : ≥95.5% (HPLC)

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple mycotoxins analysis in animal feed with LC-MS/MS: Comparison of extract dilution and immunoaffinity clean-up.
Jedziniak P, et al.
Journal of Separation Science (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service