Skip to Content
Merck

Skip To

32375

Valnemulin hydrochloride

VETRANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
CZK 6,850.00

About This Item

Empirical Formula (Hill Notation):
C31H52N2O5S · HCl
CAS Number:
Molecular Weight:
601.28
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 6,850.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

product line

VETRANAL®

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

−20°C

SMILES string

CC(C)[C@@H](N)C(NCC(C)(C)SCC(O[C@H]1[C@](C)([C@@H]2C)[C@@]3([H])[C@@](CC2)(CCC3=O)[C@@H](C)[C@H](O)[C@](C)(C=C)C1)=O)=O.Cl

InChI

1S/C31H52N2O5S.ClH/c1-10-29(8)15-22(38-23(35)16-39-28(6,7)17-33-27(37)24(32)18(2)3)30(9)19(4)11-13-31(20(5)26(29)36)14-12-21(34)25(30)31;/h10,18-20,22,24-26,36H,1,11-17,32H2,2-9H3,(H,33,37);1H/t19-,20+,22-,24-,25+,26+,29-,30+,31+;/m1./s1

InChI key

MFBPRQKHDIVLOJ-AFFLPQGKSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
323533238432379
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

application(s)

forensics and toxicology
veterinary

application(s)

forensics and toxicology
veterinary

application(s)

forensics and toxicology
veterinary

application(s)

forensics and toxicology
veterinary

format

neat

format

neat

format

neat

format

neat

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Valnemulin hydrochloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Acute Tox. 4 Oral

pictograms

Exclamation mark

signalword

Warning

hcodes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters