Skip to Content
Merck

32183

Luxabendazole-(methyl-d3)

VETRANAL®, analytical standard

Synonym(s):

2-[(Methoxy-d3-carbonyl)amino]-1H-benzimidazol-5-yl 4-fluorobenzenesulfonate, 4-Fluorobenzenesulfonic acid 2-[(methoxy-d3-carbonyl)amino]-1H-benzimidazol-5-yl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15D3H9FN3O5S
CAS Number:
Molecular Weight:
368.35
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library