Skip to Content
Merck

27488

Citric acid

puriss. p.a., ACS reagent, anhydrous, ≥99.5% (T)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
EC Number:
201-069-1
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
782061
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent, puriss. p.a.

assay

≥99.5% (T)

expl. lim.

8 %, 65 °F

ign. residue

≤0.02% (as SO4)

loss

≤0.5% loss on drying, 110 °C

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg, oxalate (C2O42-): ≤500 mg/kg, phosphate (PO43-): ≤5 mg/kg, sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤3 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤2 mg/kg, Zn: ≤5 mg/kg

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)



Still not finding the right product?

Explore all of our products under Citric acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library