Key Documents
26161
Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800
for use with Hamilton 1002 LT-LTN-RN-TLL, pkg of 1 ea
Select a Size
1 EA
CZK 944.00
CZK 944.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
CZK 944.00
About This Item
UNSPSC Code:
41122001
CZK 944.00
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE plunger tip
size
2.5 mL
feature
1002 LT-LTN--RN-TLL
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 1360-01
compatibility
for use with Hamilton 1002 LT-LTN-RN-TLL
General description
For field repair of Hamilton Series 1000, 1700 and 1800 syringes.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)