Skip to Content
Merck
All Photos(1)

Documents

25410

Sigma-Aldrich

2-Chloro-4-nitrobenzoic acid

technical, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
201.56
Beilstein:
1912836
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (HPLC)

mp

136-140 °C (lit.)
136-140 °C

solubility

methanol: soluble 0.5 g/10 mL, very faintly turbid, faintly yellow-green

SMILES string

OC(=O)c1ccc(cc1Cl)[N+]([O-])=O

InChI

1S/C7H4ClNO4/c8-6-3-4(9(12)13)1-2-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

QAYNSPOKTRVZRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-4-nitrobenzoic acid is an active pharmaceutical ingredient. Cocrystal of 2-chloro-4-nitrobenzoic acid has been prepared by Kofler hot stage contact method. Degradation of 2-chloro-4-nitrobenzoic acid by Acinetobacter sp. RKJ12 via oxygenolytic dehalogenation has been reported.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Lemmerer et al.
Journal of pharmaceutical sciences, 99(9), 4054-4071 (2010-06-25)
The active pharmaceutical ingredient 2-chloro-4-nitrobenzoic acid (2c4n) is a potentially novel therapy for immunodeficiency diseases as an anti-viral and anti-cancer agent, and exists as a dimorph in the solid state. The Kofler hot stage contact method was employed to investigate
Dhan Prakash et al.
Applied and environmental microbiology, 77(18), 6606-6613 (2011-08-02)
The organism Acinetobacter sp. RKJ12 is capable of utilizing 2-chloro-4-nitrobenzoic acid (2C4NBA) as a sole source of carbon, nitrogen, and energy. In the degradation of 2C4NBA by strain RKJ12, various metabolites were isolated and identified by a combination of chromatographic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service