Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Please contact Customer Service for Availability | CZK 2,120.00 |
About This Item
material
bevel tip stainless steel needle (point style 2), glass body
needle size
22s ga (bevel tip)
feature
removable needle RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81130
needle L
51 mm (2 in.)
volume
250 μL
compatibility
for use with Agilent/HP 1000 and 1100 LC Autosamplers
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| material bevel tip stainless steel needle (point style 2), glass body | material bevel tip stainless steel needle (point style 2), glass body | material bevel tip stainless steel needle (point style 2), clear glass body | material clear glass body, stainless steel needle (point style 2) |
| needle L 51 mm (2 in.) | needle L 51 mm (2 in.) | needle L 51 mm (2 in.) | needle L 51 mm (2 in.) |
| volume 250 μL | volume 50 μL | volume 100 μL | volume 25 μL |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename Hamilton 81130 | manufacturer/tradename Hamilton 80930 | manufacturer/tradename Hamilton 81030 | manufacturer/tradename Hamilton 80230 |
| needle size 22s ga (bevel tip) | needle size 22s ga (bevel tip) | needle size 22s ga (bevel tip) | needle size 22s ga (bevel tip) |
Still not finding the right product?
Explore all of our products under Hamilton® GASTIGHT® syringe, 1700 series, removable needle
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Skin Sens. 1
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




