20770-U
Hamilton® Syringe Cleaner
AC/DC input 120 V AC, L × W × H 6.0 in. × 1.8 in. × 2.7 in.
Synonym(s):
14-824-50
About This Item
Recommended Products
material
metal body
feature
Cleans MODIFIED MICROLITER with only heat and cleans MICROLITER™ AND GASTIGHT® syringes when a voltage rheostat is added to reduce the chamber temperature.
manufacturer/tradename
Hamilton 76610
packaging
1 ea of
L × W × H
6.0 in. × 1.8 in. × 2.7 in.
General description
Cleans Hamilton syringes and other syringes with needles up to 18 gauge (0.049 in. O.D.). Just connect the unit to your water aspirator or other vacuum source, insert the needle through the septum and, in 10-30 seconds, you′re ready for a new sample. A combination of heat (250 °C) and vacuum vaporizes and removes contaminating volatiles in a single operation.
No liquids are required with Hamilton Series 7000 syringes. For Series 700 and other conventional syringes, use a rheostat to lower the temperature (to 50 °C or below) and maintain a flow of solvent through the barrel and needle.
Instructions are included.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Syringes are precision instruments and need care. Learn to correctly inspect, use, unclog, handle, clean and store them to ensure good accuracy, reproducibility, and lifetime.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service