Skip to Content
Merck
All Photos(1)

Key Documents

16166

Sigma-Aldrich

SODOSIL RM 01

alkaline cleaner

Sign Into View Organizational & Contract Pricing

Select a Size

5 L
CZK 2,960.00
30 KG
CZK 8,370.00

CZK 2,960.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 L
CZK 2,960.00
30 KG
CZK 8,370.00

About This Item

UBA Number:
08050024
UNSPSC Code:
12352100
NACRES:
NA.22

CZK 2,960.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

pH

12-14 (20 °C, direct)

density

1.12-1.18 g/mL at 20 °C

Application

SODOSIL was used as detergent in rinsing of epoxy coated surfaces[1].

Reconstitution

The best cleaning results are achieved with = 2−20% for normal contamination. The cleaning time = 2−24 h depends on the degree of contamination and on the bath temperature. The action of ultra-sonic waves accelerates the cleaning operation considerably as does an increase in temperature. The cleaning solution can be prepared with tap water. The use of demineralisied water results in more optimum utilisation of baths.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Aspenes et al.
Journal of colloid and interface science, 343(2), 529-536 (2009-12-29)
The mechanisms by which hydrates deposit in a petroleum production line are related to pipeline surface properties, fluid composition and properties, and water cut. In this work, adhesion forces between cyclopentane hydrates and solid surfaces were investigated as a function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service