Skip to Content
Merck
All Photos(1)

Documents

16140

Sigma-Aldrich

2-Bromoethanol

purum, ≥95.0% (GC)

Synonym(s):

Ethylene bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2OH
CAS Number:
Molecular Weight:
124.96
Beilstein:
878140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.3 (vs air)

vapor pressure

2.4 mmHg ( 20 °C)

grade

purum

Assay

≥95.0% (GC)

refractive index

n20/D 1.492 (lit.)
n20/D 1.494

bp

56-57 °C/20 mmHg (lit.)

density

1.763 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCBr

InChI

1S/C2H5BrO/c3-1-2-4/h4H,1-2H2

InChI key

LDLCZOVUSADOIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to brown on storage

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Khan et al.
Biochemical pharmacology, 45(2), 439-447 (1993-01-26)
The cytotoxicity of dibromoalkanes to isolated hepatocytes was proportional to the dibromoalkane concentration and increasing chain length of the dibromoalkane (C2-C6). The rapid hepatocyte glutathione (GSH) depletion which occurred upon addition of the dibromoalkanes was also dependent on the concentration
W Van Rillaer et al.
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 189(1), 21-25 (1989-07-01)
This study describes a headspace gas-chromatographic method for the quantitative determination of inorganic bromide in vegetables. Different parameters influencing the reaction of the bromide ion into 2-bromoethanol have been investigated. The analysis of 2-bromoethanol is performed by the headspace--electron capture--gas
A R Jones et al.
Xenobiotica; the fate of foreign compounds in biological systems, 11(11), 763-770 (1981-11-01)
1. The metabolism of 2-bromo[U-14C]ethanol an [U-14C]ethylene oxide has been studied in the rat. 2. As both compounds give rise to similar amounts of two urinary metabolites, identified as S-(2-hydroxyethyl)cysteine and N-acetyl-S-(2-hydroxyethyl)cysteine, it is proposed that 2-bromoethanol is converted into
H Dunkelberg
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale B, Hygiene, 177(3-4), 269-281 (1983-04-01)
The compounds 2-chloroethanol and 2-bromoethanol were administered once weekly subcutaneously to groups of 100 female NMRI mice, in the case of 2-chloroethanol in 3 dosages (3.0, 1.0 and 0.3 mg, single dosage per mouse) and 2-bromoethanol in 2 dosages (1.0
Caroline C Womack et al.
The journal of physical chemistry. A, 115(51), 14559-14569 (2011-11-11)
This work characterizes the internal energy distribution of the CD(2)CD(2)OH radical formed via photodissociation of 2-bromoethanol-d(4). The CD(2)CD(2)OH radical is the first radical adduct in the addition of the hydroxyl radical to C(2)D(4) and the product branching of the OH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service