Skip to Content
Merck

Skip To

15747

Silicon Standard for ICP

TraceCERT®, 1 g/L Si in NaOH (nominal concentration)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 1,250.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

CZK 1,250.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

concentration

1 g/L Si in NaOH (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

50 ng/g of Silicon standard may be used to improve maximum signal-to-background intensity ratios (SBR) during High-resolution inductively coupled plasma mass spectrometry (HR-ICP-MS).

Analysis Note

1 g/L Si in 2% sodium hydroxide, prepared with high purity Si, NaOH and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
167341802119078
technique(s)

ICP: suitable

technique(s)

ICP: suitable

technique(s)

ICP: suitable

technique(s)

ICP: suitable

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

format

single component solution

format

single component solution

format

single component solution

format

single component solution

product line

TraceCERT®

product line

TraceCERT®

product line

TraceCERT®

product line

TraceCERT®

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

concentration

1 g/L Si in NaOH (nominal concentration)

concentration

1 g/L Ce in nitric acid (nominal concentration)

concentration

1 g/L S in H2O (nominal concentration)

concentration

1 g/L Pt in hydrochloric acid (nominal concentration)


Still not finding the right product?

Explore all of our products under Silicon Standard for ICP


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters