Skip to Content
Merck
All Photos(1)

Key Documents

14121

Millipore

Lactic Acid Supplement, modified

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

5 AMP
CZK 4,410.00

CZK 4,410.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 AMP
CZK 4,410.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

CZK 4,410.00


Please contact Customer Service for Availability

Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

bacteria

General description

(per vial, sufficient for 500 mL medium):
Sorbitan mono-oleate 5.0 mL, 2-phenyl ethanol 1.5 g, Amphotericin 2.5 mg

Application

Lactic acid could be used in the culture medium for the mouse testis cells, used for the long-term spermatogonial stem cell culture, in order to study spermatogenesis mechanism that has future benefits of being used in developing new technology in transgenesis or medicine.[1]

Components

(per vial, sufficient for 500 mL medium):
Sorbitan mono-oleate 5.0 mL, 2-phenyl ethanol 1.5 g, Amphotericin 2.5 mg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mito Kanatsu-Shinohara et al.
Biology of reproduction, 69(2), 612-616 (2003-04-18)
Spermatogenesis is a complex process that originates in a small population of spermatogonial stem cells. Here we report the in vitro culture of spermatogonial stem cells that proliferate for long periods of time. In the presence of glial cell line-derived

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service