Skip to Content
Merck

13490

N-Isopropylbenzylamine

purum, ≥97.0% (GC)

Synonym(s):

N-Benzylisopropylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2NHCH(CH3)2
CAS Number:
Molecular Weight:
149.23
EC Number:
203-067-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
2638437
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.502

bp

200 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CC(C)NCc1ccccc1

InChI

1S/C10H15N/c1-9(2)11-8-10-6-4-3-5-7-10/h3-7,9,11H,8H2,1-2H3

InChI key

LYBKPDDZTNUNNM-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under N-Isopropylbenzylamine


signalword

Warning

pictograms

Exclamation mark

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library