Skip to Content
Merck
All Photos(1)

Key Documents

09782

Sigma-Aldrich

(1R,4S)-(+)-4-(Boc-amino)-2-cyclopentene-1-carboxylic acid

purum, ≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO4
CAS Number:
Molecular Weight:
227.26
Beilstein:
6177417
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (HPLC)

form

powder with small lumps

optical activity

[α]20/D +48.5±2°, c = 1% in methanol

color

white to faintly beige

mp

151-155 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@H]1C[C@H](C=C1)C(O)=O

InChI

1S/C11H17NO4/c1-11(2,3)16-10(15)12-8-5-4-7(6-8)9(13)14/h4-5,7-8H,6H2,1-3H3,(H,12,15)(H,13,14)/t7-,8+/m0/s1

InChI key

WOUNTSATDZJBLP-JGVFFNPUSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunjie Xia et al.
Cellular and molecular gastroenterology and hepatology, 12(3), 1121-1143 (2021-05-08)
Identifying novel and actionable targets in hepatocellular carcinoma (HCC) remains an unmet medical need. TAK1 was originally identified as a transforming growth factor-β-activated kinase and was further proved to phosphorylate and activate numerous downstream targets and promote cancer progression. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service