Skip to Content
Merck

Skip To

09171

mono-(2-Ethyl-5-hydroxyhexyl) phthalate, mixture of diastereomers

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1-(2-ethyl-5-hydroxyhexyl) ester, Phthalic acid mono-(2-ethyl-5-hydroxyhexyl) ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Please contact Customer Service for Availability
CZK 21,700.00

About This Item

Empirical Formula (Hill Notation):
C16H22O5
CAS Number:
Molecular Weight:
294.34
UNSPSC Code:
77101502
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
9208635

CZK 21,700.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCC(CC)CCC(C)O)C1=CC=CC=C1C(O)=O

InChI

1S/C16H22O5/c1-3-12(9-8-11(2)17)10-21-16(20)14-7-5-4-6-13(14)15(18)19/h4-7,11-12,17H,3,8-10H2,1-2H3,(H,18,19)

InChI key

RYPQSGURZSTFSX-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8013742446SMB00941
application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

-

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

-

grade

analytical standard

grade

-

assay

≥97.0% (HPLC)

assay

≥97.0% (GC)

assay

≥97.0% (HPLC)

assay

≥95%

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters