Skip to Content
Merck
All Photos(1)

Key Documents

08986

Millipore

Staphylo Monotec test kit

suitable for microbiology

Synonym(s):

Monostaph test kit, Staphylococcus Monotec test kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621

shelf life

limited shelf life, expiry date on the label

Quality Level

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Staphylococcus spp.

General description

Staphylo Monotec test kit is a new rapid agglutination test for differentiation between Staphylococcus aureus and other Staphylococcus. This method has been evaluated for clinical specimens and food material. With this kit coagulase and Protein A on S. aureus can be detected in one step.

Components

1 × Test reagent (Cat. No. 88786; vial with red cap), 5 ml
1x Control reagent (Cat. No. 69187; vial with white cap), 5 ml
40 × Analysis cards
(contains reagent for more than 100 tests)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service