Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C5H5N3O2
CAS Number:
Molecular Weight:
139.11
EC Number:
224-145-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
120353
MDL number:
Pricing and availability is not currently available.
grade
technical
assay
≥95.0% (NT)
mp
186-188 °C (lit.)
SMILES string
Nc1ccc(cn1)[N+]([O-])=O
InChI
1S/C5H5N3O2/c6-5-2-1-4(3-7-5)8(9)10/h1-3H,(H2,6,7)
InChI key
UGSBCCAHDVCHGI-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
435.2 °F
flash_point_c
224 °C
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Danila V Zimenkov et al.
Journal of clinical microbiology, 53(4), 1103-1114 (2015-01-23)
In addition to the obligatory pathogenic species of the Mycobacterium tuberculosis complex and Mycobacterium leprae, the genus Mycobacterium also includes conditionally pathogenic species that in rare cases can lead to the development of nontuberculous mycobacterial diseases. Because tuberculosis and mycobacteriosis
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8220930100 | 04022536483948 |
| 537705-18L-CS | 04061837714931 |
| 8220939046 | 04027269995898 |
| 8220931000 | 04022536483955 |
| 537705-20L | 04061837714948 |
| 537705-1L | 04061832565897 |
