Skip to Content
Merck

08158

6-Amino-4-hydroxy-2-naphthalenesulfonic acid

technical, ≥90% (T)

Synonym(s):

7-Amino-1-naphthol-3-sulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H9NO4S
CAS Number:
Molecular Weight:
239.25
EC Number:
202-000-8
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1821283
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical

assay

≥90% (T)

SMILES string

Nc1ccc2cc(cc(O)c2c1)S(O)(=O)=O

InChI

1S/C10H9NO4S/c11-7-2-1-6-3-8(16(13,14)15)5-10(12)9(6)4-7/h1-5,12H,11H2,(H,13,14,15)

InChI key

HBZVNWNSRNTWPS-UHFFFAOYSA-N



pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sanjit Saha et al.
Physical chemistry chemical physics : PCCP, 19(42), 28588-28595 (2017-10-19)
The surface electronic properties of graphene oxide (GO) were modified through reduction and functionalization. Non-covalent functionalization was found to be superior compared to covalent functionalization due to the formation of few-layer graphene with a low defect content and average crystalline



Global Trade Item Number

SKUGTIN
822203050004022536486109
822203010004022536486093
H7408-100G04061833803776
445312-25G04061832283784
445312-5G04061832283791
H7408-500G04061833803783