Skip to Content
Merck

08123

1-Decanesulfonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)9SO2Cl
CAS Number:
Molecular Weight:
240.79
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

CCCCCCCCCCS(Cl)(=O)=O

InChI

1S/C10H21ClO2S/c1-2-3-4-5-6-7-8-9-10-14(11,12)13/h2-10H2,1H3

InChI key

WCKKWABZCCKDTN-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library