Skip to Content
Merck
All Photos(1)

Key Documents

07752

Supelco

Metolachlor-(2-ethyl-6-methylphenyl-d11)

PESTANAL®, analytical standard

Synonym(s):

Metolachlor-d11

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 6,880.00

CZK 6,880.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 6,880.00

About This Item

Empirical Formula (Hill Notation):
C15D11H11ClNO2
CAS Number:
Molecular Weight:
294.86
UNSPSC Code:
77101502
NACRES:
NA.24

CZK 6,880.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥98 atom % D

product line

PESTANAL®

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c(c(N(C(C)COC)C(=O)CCl)c(c1[2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3/i1D3,2D3,5D2,6D,7D,8D

InChI key

WVQBLGZPHOPPFO-ZPOKHESBSA-N

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service