Skip to Content
Merck

07267

Protopine

analytical standard

Synonym(s):

4,6,7,14-Tetrahydro-5-methylbis[1,3]benzodioxolo[4,5-c:5′,6′-g]azecin-13(5H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H19NO5
CAS Number:
Molecular Weight:
353.37
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

application(s)

food and beverages

format

neat

SMILES string

O=C1CC2=C(C(OCO3)=C3C=C2)CN(C)CCC4=CC5=C(OCO5)C=C41

InChI

1S/C20H19NO5/c1-21-5-4-13-7-18-19(25-10-24-18)8-14(13)16(22)6-12-2-3-17-20(15(12)9-21)26-11-23-17/h2-3,7-8H,4-6,9-11H2,1H3

InChI key

GPTFURBXHJWNHR-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library