Skip to Content
Merck

05945

Allyldimethylsilane

purum, ≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH2=CHCH2Si(CH3)2H
CAS Number:
Molecular Weight:
100.23
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
1734304
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (GC)

bp

69 °C (lit.)

density

0.710 g/mL at 20 °C (lit.)





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library