Skip to Content
Merck
All Photos(1)

Documents

05320590

p-Anisaldehyde

primary reference standard

Synonym(s):

4-Methoxybenzaldehyde, Aubépine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
471382
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

vapor density

4.7 (vs air)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.573 (lit.)

bp

248 °C (lit.)

mp

−1 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

application(s)

food and beverages

storage temp.

−20°C

SMILES string

[H]C(=O)c1ccc(OC)cc1

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service