Skip to Content
Merck

Skip To

04887

Redox standard

124 mV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Please contact Customer Service for Availability
CZK 4,270.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

CZK 4,270.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

description

redox potential: 124 mV ±5 mV, 25°C

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

2-8°C

General description

This Redox standard has a potential of 124 mV ±5 mV. It refers to the potential of the platinum electrode versus the Ag/AgCl reference electrode in 3M potassium chloride.

Application

Redox standard acts as a control standard rather than a calibration standard which controls the functionality of the sensing and reference electrodes.

Features and Benefits

  • Broad range of values
  • Very high specifications (±5mV)
  • Detailed safety data sheet (SDS) available
  • Certified with proven verifiable accuracy and uncertainty of measurement
  • Guaranteed stability throughout its entire shelf life

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
877837885580138
format

mixture

format

mixture

format

mixture

format

mixture

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

application(s)

environmental
pharmaceutical

application(s)

environmental
pharmaceutical

application(s)

environmental
pharmaceutical

application(s)

environmental
pharmaceutical

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Redox standard


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters