Skip to Content
Merck

04855

(S)-2-Oxiranylanisole

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(S)-α-(2-Oxiranyl)anisole, (S)-Glycidyl phenyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352005
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥97.0% (sum of enantiomers, GC)

SMILES string

C1O[C@@H]1COc2ccccc2

InChI

1S/C9H10O2/c1-2-4-8(5-3-1)10-6-9-7-11-9/h1-5,9H,6-7H2/t9-/m1/s1

InChI key

FQYUMYWMJTYZTK-SECBINFHSA-N

General description

The epoxide hydrolase isolated from Bacillus megaterium ECU1001 can preferentially hydrolyze the (R)-enantiomer of racemic phenyl glycidyl ether to yield optically pure (S)-phenyl glycidyl ether.


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

159.8 °F

flash_point_c

71 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Biocatalytic preparation of (S)-phenyl glycidyl ether using newly isolated Bacillus megaterium ECU1001.
Journal of Molecular Catalysis. B, Enzymatic, 13(4), 61-68 (2001)



Global Trade Item Number

SKUGTIN
04855-5G-F04061838635815