Skip to Content
Merck

04584

Rappaport Vassiliadis Broth acc. to DIN EN ISO 6579:2002

suitable for microbiology

Synonym(s):

Rappaport Vassiliadis Soya Broth

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41171606
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

shelf life

limited shelf life, expiry date on the label

composition

dipotassium phosphate, 0.18 g/L , magnesium chloride, 13.58 g/L , malachite green, 0.036 g/L , monopotassium phosphate, 1.26 g/L , sodium chloride, 7.2 g/L , soy peptone, 4.50 g/L

final pH

5.2±0.2 (25 °C)

application(s)

environmental
food and beverages, microbiology

suitability

selective for Salmonella spp.

General description

Liquid medium for the selective enrichment of Salmonella in foodstuffs and other materials. The Rappaport Vassiliadis medium complies with recommendations of the APHA for the examinations of food.


Storage Class

13 - Non Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library