Skip to Content
Merck

04072

Fmoc-7-Ahp-OH

≥98.0% (HPLC)

Synonym(s):

7-(Fmoc-amino)enanthic acid, 7-(Fmoc-amino)heptanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H25NO4
CAS Number:
Molecular Weight:
367.44
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (HPLC)

form

powder with small lumps

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C22H25NO4/c24-21(25)13-3-1-2-8-14-23-22(26)27-15-20-18-11-6-4-9-16(18)17-10-5-7-12-19(17)20/h4-7,9-12,20H,1-3,8,13-15H2,(H,23,26)(H,24,25)

InChI key

FRPYXTFBPCYALT-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library