Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C23H19NO4
CAS Number:
Molecular Weight:
373.40
UNSPSC Code:
12352200
NACRES:
NA.26
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
7391207
assay
≥98.0%
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
color
white to almost white
mp
224-228 °C
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
SMILES string
OC(=O)c1ccc(CNC(=O)OCC2c3ccccc3-c4ccccc24)cc1
InChI
1S/C23H19NO4/c25-22(26)16-11-9-15(10-12-16)13-24-23(27)28-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21H,13-14H2,(H,24,27)(H,25,26)
InChI key
JRHUROPSUJVMNH-UHFFFAOYSA-N
Other Notes
Rigid linker employed in preparing Rink amide resins; Prep. of aminomethyl resin by coupling with Wang resin.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.