Skip to Content
Merck
All Photos(1)

Key Documents

03395

Sigma-Aldrich

Poly(ethylene glycol)

tested according to Ph. Eur., 1,000

Synonym(s):

Macrogol 1000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

Agency

tested according to Ph. Eur.

Quality Level

form

waxy solid

mol wt

950-1050

solubility

aliphatic hydrocarbons: slightly soluble
organic solvents: soluble

application(s)

pharmaceutical (small molecule)

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) is a chemical fusogen, which mediates the premature chromosome condensation induction in human and rat lymphocytes as well as in rodent spleen cells, and spleen and whole body cells of newborn mice.[1]

Biochem/physiol Actions

Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water. It enhances the refolding of extracted proteins. This enables characterization of crystallized proteins.[2][3]

Preparation Note

Poly(ethylene glycol) dissolves in many organic solvents and is readily soluble in aromatic hydrocarbons. It also slightly dissolves in aliphatic hydrocarbons.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G E Pantelias et al.
Somatic cell genetics, 9(5), 533-547 (1983-09-01)
Even though polyethylene glycol (PEG) has been shown to be a potent fusogen, it has not been widely exploited as an alternative to the Sendai virus for premature chromosome condensation (PCC) induction. A simple, rapid, and reproducible PEG protocol for
J L Cleland et al.
Bio/technology (Nature Publishing Company), 10(9), 1013-1019 (1992-09-01)
Previous studies on the refolding of recombinant bovine carbonic anhydrase B (CAB) indicated that polyethylene glycol (PEG) significantly enhanced the recovery of active protein by reducing aggregation. To further test the ability of PEG to enhance refolding, three recombinant human
J L Cleland et al.
The Journal of biological chemistry, 267(19), 13327-13334 (1992-07-05)
Polyethylene glycol (PEG) inhibited aggregation during refolding of bovine carbonic anhydrase B (CAB) through the formation of a nonassociating PEG-intermediate complex. Stoichiometric concentrations of PEG were required for complete recovery of active protein during refolding at aggregating conditions. For example
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service