Skip to Content
Merck

03340

Chloroethane

purum, ≥98.0%

Synonym(s):

Ethyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51
UNSPSC Code:
12142100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
200-830-5
Beilstein/REAXYS Number:
1730751
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

2.22 (vs air)

vapor pressure

19.2 psi ( 20 °C)

grade

purum

assay

≥98.0%

impurities

diethylether, water

bp

12.3 °C (lit.)

mp

−139 °C (lit.)

density

0.921 g/mL at 0-4 °C (lit.)

SMILES string

CCCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

Packaging

Pressure tin with 100 mL (net ~89 g)

Other Notes

Sales restrictions may apply


Still not finding the right product?

Explore all of our products under Chloroethane


signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - Repr. 1B

Storage Class

2A - Gases

wgk

WGK 2

flash_point_f

-58.0 °F - closed cup

flash_point_c

-50.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Y-G Im et al.
Journal of oral rehabilitation, 39(12), 931-940 (2012-09-22)
The aim of this study was to compare the effects of three cryotherapeutic modalities (ethyl chloride spraying, ice block rubbing and cold gel packing) on facial skin temperature. Thirty healthy volunteers (15 men, 15 women; mean age, 29·4 ± 3·2 years) participated in
Robert N Richards
Journal of cutaneous medicine and surgery, 13(5), 253-256 (2009-09-23)
Botulinum toxin injections are effective in the treatment of palmar and plantar hyperhidrosis, but discomfort has limited its use. To study the use of ethyl chloride medium-stream spray in reducing injection discomfort. We used ethyl chloride medium-stream spray, in conjunction
Lindsey L Cohen et al.
The Clinical journal of pain, 25(6), 490-494 (2009-06-23)
The purpose of this study was to evaluate the effectiveness of vapocoolant for preschoolers' immunization injection pain relief. Fifty-seven 4 to 6-year-old children were randomized into vapocoolant alone or typical care conditions. Pain was measured at the baseline and at



Global Trade Item Number

SKUGTIN
821461001004022536477176
821461010004027269073749