Skip to Content
Merck

00733

Ethyl decanoate

analytical standard

Synonym(s):

Capric acid ethyl ester, Ethyl caprate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)8COOC2H5
CAS Number:
Molecular Weight:
200.32
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-761-9
Beilstein/REAXYS Number:
1762128
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

vapor density

6.9 (vs air)

assay

≥99.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

impurities

≤0.3% water

refractive index

n20/D 1.423-1.427, n20/D 1.425 (lit.)

bp

245 °C (lit.)

density

0.864 g/cm3

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

ester

SMILES string

CCCCCCCCCC(=O)OCC

InChI

1S/C12H24O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h3-11H2,1-2H3

InChI key

RGXWDWUGBIJHDO-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Ethyl decanoate


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library