Skip to Content
Merck
All Photos(1)

Key Documents

00496

Sigma-Aldrich

cis-4-Hepten-1-ol

≥95.0%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CZK 875.00

CZK 875.00

List PriceCZK 1,250.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
CZK 875.00

About This Item

Empirical Formula (Hill Notation):
C7H14O
CAS Number:
Molecular Weight:
114.19
Beilstein:
1840300
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020352
PubChem Substance ID:
NACRES:
NA.22

CZK 875.00

List PriceCZK 1,250.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0%

refractive index

n20/D 1.445

bp

78 °C/23 mmHg (lit.)

density

0.850 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CC\C=C/CCCO

InChI

1S/C7H14O/c1-2-3-4-5-6-7-8/h3-4,8H,2,5-7H2,1H3/b4-3-

InChI key

CUKAXHVLXKIPKF-ARJAWSKDSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service