Skip to Content
Merck
All Photos(1)

Key Documents

28-6033

Sigma-Aldrich

Sulfuric acid (1+1)

~64.0 % (w/w) in H2O

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

~64.0 % (w/w) in H2O

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengcheng Li et al.
Bioresource technology, 130, 629-637 (2013-01-22)
Xylans are the major hemicelluloses in grasses, but their effects on biomass saccharification remain unclear. In this study, we examined the 79 representative Miscanthus accessions that displayed a diverse cell wall composition and varied biomass digestibility. Correlation analysis showed that
Michael Schinhammer et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1882-1893 (2013-03-19)
Biodegradable metals offer great potential in circumventing the long-term risks and side effects of medical implants. Austenitic Fe-Mn-C-Pd alloys feature a well-balanced combination of high strength and considerable ductility which make them attractive for use as degradable implant material. The
M K Mohamad Haafiz et al.
Carbohydrate polymers, 93(2), 628-634 (2013-03-19)
In this work, we successfully isolated microcrystalline cellulose (MCC) from oil palm empty fruit bunch (OPEFB) fiber-total chlorine free (TCF) pulp using acid hydrolysis method. TCF pulp bleaching carried out using an oxygen-ozone-hydrogen peroxide bleaching sequence. Fourier transform infrared (FT-IR)
J Gamble et al.
Environmental research, 35(1), 30-52 (1984-10-01)
The effects of long-term exposure to sulfuric acid mist on the teeth and respiratory system were studied in 248 workers in five plants manufacturing lead acid batteries. The prevalence of cough, phlegm, dyspnea, and wheezing as determined by questionnaire were
Talita M Lacerda et al.
Carbohydrate polymers, 93(1), 347-356 (2013-03-08)
The influence of sulfuric acid concentration (H2SO4 5-25%, 100°C), crystallinity and fibers size on the hydrolysis reaction of sisal pulps were investigated, with the goal of evaluating both the liquor composition, as an important step in the production of bioethanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service