Skip to Content
Merck
All Photos(1)

Key Documents

28-4980

Sigma-Aldrich

Starch, soluble

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1

InChI key

GUBGYTABKSRVRQ-ASMJPISFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murali Krishna Chimata et al.
Biotechnology research international, 2011, 323891-323891 (2011-10-07)
The production of extracellular amylase was investigated employing our laboratory isolate, Aspergillus niger sp. MK 07 and effect of process variables on enzyme production, was studied in a fermentor. It was found that amylase production was maximum when the fermentor
M Ohnishi et al.
Carbohydrate research, 161(2), 257-263 (1987-04-01)
Acid hydrolyzates of waxy-maize starch were separated to give Fractions I, II, and III [T. Watanabe, and D. French, Carbohydr. Res., 84 (1980) 115-123]. Watanabe and French suggested that Fraction II, which contains approximately 25 D-glucose residues including an alpha-D-(1----6)-linked
J L Jane et al.
Carbohydrate research, 140(1), 21-35 (1985-07-01)
Amylose (average d.p. 1000) and amylodextrin (average d.p. 25) have identical 13C-n.m.r. spectra, except for some minor signals from the small amount of alpha-1----6 branch linkages present in amylodextrin. Amylodextrin can be obtained as stable solutions in much higher concentrations
J Van der Veen et al.
Pharmaceutical research, 11(4), 499-502 (1994-04-01)
Amylodextrin is a linear dextrin and can be produced by enzymatic hydrolysis of the alpha-1,6 glycosidic bonds of amylopectin. Tablets compacted from pure amylodextrin showed good binding properties and did not disintegrate in aqueous media. Extended and decreasing drug release
J van der Veen et al.
Pharmaceutical research, 11(3), 384-387 (1994-03-01)
Amylodextrin is a suitable excipient for the design of solid controlled-release systems. The release of paracetamol from tablets containing 30% drug and 70% amylodextrin was studied in vitro and in vivo. In vitro dissolution profiles showed almost-constant drug release rates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service