Skip to Content
Merck
All Photos(1)

Key Documents

28-4840

Sigma-Aldrich

Sorbitan monooleate

SAJ first grade

Synonym(s):

Span® 80

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H44O6
CAS Number:
Molecular Weight:
428.60
Beilstein:
8172514
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
SAJ first grade

grade

SAJ first grade

form

liquid

availability

available only in Japan

refractive index

n20/D 1.48 (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

[H][C@@](O)(COC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@H](O)[C@H]1O

InChI

1S/C24H44O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h9-10,20-21,23-26,28H,2-8,11-19H2,1H3/b10-9-/t20-,21+,23+,24+/m0/s1

InChI key

NWGKJDSIEKMTRX-AAZCQSIUSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

485.6 °F

Flash Point(C)

252 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faiyaz Shakeel et al.
Drug development and industrial pharmacy, 40(9), 1240-1245 (2013-07-11)
Abstract For the development of an effective self-nanoemulsifying drug delivery system (SNEDDS) of poorly soluble drugs, the knowledge of the solubility in its oil phase and SNEDDS are one of the most important factors to avoid possibility of drug to
Hoang Hirschberg et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 46(1-2), 1-7 (2012-02-15)
In the search for an optimal approach for the transcutaneous immunization (TCI) of hepatitis B surface antigen (HBsAg), two vesicle formulations, L595 vesicles (composed of sucrose-laurate ester and octaoxyethylene-laurate ester) and sPC vesicles (composed of soybean-phosphatidylcholine and Span-80) were prepared
Daniela S Bernardi et al.
Journal of nanobiotechnology, 9, 44-44 (2011-09-29)
Nanoemulsions have practical application in a multitude of commercial areas, such as the chemical, pharmaceutical and cosmetic industries. Cosmetic industries use rice bran oil in sunscreen formulations, anti ageing products and in treatments for skin diseases. The aim of this
Geneviève LeBel et al.
Applied and environmental microbiology, 80(17), 5484-5492 (2014-06-29)
Streptococcus suis serotype 2 is known to cause severe infections (meningitis, endocarditis, and septicemia) in pigs and is considered an emerging zoonotic agent. Antibiotics have long been used in the swine industry for disease treatment/prevention and growth promoters. This pattern
Maroof Alam et al.
Nanomedicine : nanotechnology, biology, and medicine, 9(2), 247-256 (2012-08-04)
In the current study, a novel niosome based formulation of diallyl disulfide (DADS) was evaluated for its potential to treat disseminated candidiasis in mouse model. Among various non-ionic surfactants tested, niosome formulation prepared using Span 80 was found to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service