Skip to Content
Merck
All Photos(1)

Key Documents

28-3840

Sigma-Aldrich

Sodium phosphate tribasic dodecahydrate

SAJ first grade, 98.0-102.0%

Synonym(s):

TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3PO4 · 12H2O
CAS Number:
Molecular Weight:
380.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
98.0-102.0%
grade:
SAJ first grade
form:
crystalline

grade

SAJ first grade

Assay

98.0-102.0%

form

crystalline

availability

available only in Japan

pH

13 (25 °C, 190.1 g/L)

density

1.62 g/mL at 25 °C

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

InChI

1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3

InChI key

ASTWEMOBIXQPPV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gisela Ferraro et al.
Journal of experimental botany, 66(11), 3381-3389 (2015-04-17)
Glutamate (Glu) is a taste enhancer that contributes to the characteristic flavour of foods. In fruit of tomato (Solanum lycopersicum L.), the Glu content increases dramatically during the ripening process, becoming the most abundant free amino acid when the fruit
Joshua C Cheng et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(26), 9689-9700 (2015-07-03)
Temporal summation of pain (TSP), the perception of increasingly greater pain evoked by repetitive noxious stimuli, is highly variable between individuals. Individuals with facilitated pain processing and/or reduced pain-modulatory capabilities are regarded as pronociceptive, whereas individuals with reduced pain processing
Santosh Lamichhane et al.
Analytical chemistry, 87(12), 5930-5937 (2015-05-20)
Metabolomic analyses of fecal material are gaining increasing attention because the gut microbial ecology and activity have an impact on the human phenotype and regulate host metabolism. Sample preparation is a crucial step, and in this study, we recommend a
Jie Mu et al.
Seizure, 23(7), 553-559 (2014-05-24)
To evaluate the value of magnetoencephalography (MEG) source localization in localization of epileptic activities and predicting surgical outcome in frontal lobe epilepsies (FLE). Forty-six patients with presurgical MEG evaluation and intractable FLE surgery (28 male patients) were analyzed retrospectively with
Claudio Pulito et al.
Oncotarget, 6(20), 18134-18150 (2015-07-03)
Malignant pleural mesothelioma is a poorly treated neoplasia arising from the pleural mesothelial lining. Here we document that the leaf extract of Cynara scolymus exerts broad antitumoral effects both in vitro and in vivo on mesothelioma cell lines. We found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service