Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
EC Number:
201-176-3
UNSPSC Code:
12352106
PubChem Substance ID:
Beilstein/REAXYS Number:
506071
MDL number:
grade
SAJ first grade
vapor density
2.55 (vs air)
vapor pressure
2.4 mmHg ( 20 °C)
assay
≥98.0%
autoignition temp.
955 °F
expl. lim.
12.1 %
availability
available only in Japan
refractive index
n20/D 1.386 (lit.)
bp
141 °C (lit.)
mp
−24-−23 °C (lit.)
density
0.993 g/mL at 25 °C (lit.)
SMILES string
CCC(O)=O
InChI
1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)
InChI key
XBDQKXXYIPTUBI-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Propionic acid
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
129.2 °F - closed cup
flash_point_c
54 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


