Skip to Content
Merck
All Photos(1)

Key Documents

24-5570

Sigma-Aldrich

Potassium stearate

CP

Synonym(s):

Octadecanoic acid potassium salt, Stearic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOK
Molecular Weight:
322.57
UNSPSC Code:
12352300

grade

CP

form

solid

availability

available only in Japan

storage temp.

15-25°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerardo Alvarez-Rivera et al.
Journal of chromatography. A, 1440, 37-44 (2016-03-08)
The stability and photochemical transformations of cosmetic preservatives in topical applications exposed to UV-light is a serious but poorly understood problem. In this study, a high throughput extraction and selective method based on pressurized liquid extraction (PLE) coupled to gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service