Skip to Content
Merck
All Photos(1)

Key Documents

18-1760

Sigma-Aldrich

Luminol

SAJ special grade

Synonym(s):

3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
383929
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ special grade

form

powder

availability

available only in Japan

mp

>300 °C (lit.)

SMILES string

Nc1cccc2C(=O)NNC(=O)c12

InChI

1S/C8H7N3O2/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12/h1-3H,9H2,(H,10,12)(H,11,13)

InChI key

HWYHZTIRURJOHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used for chemiluminescence analysis of, for example, metal cations, blood, and glucococorticoids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reference values of reactive oxygen species in seminal ejaculates using chemiluminescence assay
Agarwal A, et al.
Journal of Assisted Reproduction and Genetics, 32(12), 1721-1729 (2015)
Luminol-based chemiluminescent signals: clinical and non-clinical application and future uses
Khan P, et al.
Applied Biochemistry and Biotechnology, 173(2), 333-355 (2014)
Christophe A Marquette et al.
Analytical and bioanalytical chemistry, 385(3), 546-554 (2006-05-23)
This critical review discusses the results published between 2000 and 2005 on the development of analytical systems based on the luminol chemiluminescent and electrochemiluminescent reactions. An increasing number of non-specific detection systems based on the enhancing, inhibiting or catalysing effect
Feng-Ming Hsu et al.
Scientific reports, 5, 10291-10291 (2015-05-20)
Esophageal cancer patients with pathological complete response (pCR) to neoadjuvant chemoradiation (CRT) have favorable outcomes. Currently, there was no reliable biomarker predicting the response to CRT. Perioperative circulating mRNA may be associated with prognosis, but its application for predicting treatment
Fuan Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(50), 16030-16036 (2012-10-20)
The Cu(2+)-dependent ligation DNAzyme is implemented as a biocatalyst for the colorimetric or chemiluminescence detection of Cu(2+) ions, Hg(2+) ions, or cocaine. These sensing platforms are based on the structural tailoring of the sequence of the Cu(2+)-dependent ligation DNAzyme for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service