Skip to Content
Merck
All Photos(1)

Key Documents

18-1440

Sigma-Aldrich

Lithium chloride

SAJ special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

6

mp

605 °C (lit.)

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Di Maio et al.
EvoDevo, 6, 17-17 (2015-07-15)
Wnt signaling is one of the earliest and most highly conserved regulatory pathways for the establishment of the body axes during regeneration and early development. In regeneration, body axes determination occurs independently of tissue rearrangement and early developmental cues. Modulation
A M Petrosyan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 105, 623-625 (2013-02-07)
We argue that the recently reported crystals "L-threonine formate" as well as "L-alanine lithium chloride" and "bis L-alanine lithium chloride" actually are the well-known crystals L-threonine and L-alanine, respectively.
A Abu-Baker et al.
Cell death & disease, 4, e821-e821 (2013-10-05)
Expansion of polyalanine tracts causes at least nine inherited human diseases. Among these, a polyalanine tract expansion in the poly (A)-binding protein nuclear 1 (expPABPN1) causes oculopharyngeal muscular dystrophy (OPMD). So far, there is no treatment for OPMD patients. Developing
Aitor González et al.
PloS one, 8(1), e53323-e53323 (2013-01-18)
The mouse segmentation is established from somites, which are iteratively induced every two hours from the presomitic mesoderm (PSM) by a system known as the segmentation clock. A crucial component of the segmentation clock is the gene Hes7, which is
Karla M Acevedo et al.
The Journal of biological chemistry, 289(16), 11007-11019 (2014-03-13)
Amyloid precursor protein (APP) undergoes post-translational modification, including O- and N-glycosylation, ubiquitination, and phosphorylation as it traffics through the secretory pathway. We have previously reported that copper promotes a change in the cellular localization of APP. We now report that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service