Skip to Content
Merck

Skip To

07-4430

N,N-Dimethylformamide

JIS special grade, ≥99.5%

Synonym(s):

DMF, NSC 5356

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
PubChem Substance ID:
UNSPSC Code:
12352111
Beilstein/REAXYS Number:
605365
MDL number:
Assay:
≥99.5%
Grade:
JIS special grade
Bp:
153 °C (lit.)
Vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

JIS special grade

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

assay

≥99.5%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.430 (lit.)

pH

6.7

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Application

Solvent for many hydrophobic organic compounds.


Still not finding the right product?

Explore all of our products under N,N-Dimethylformamide


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

136.4 °F

flash_point_c

58 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

68-12-2

CAS No.

68-12-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters