Skip to Content
Merck
All Photos(1)

Documents

07-2920

Sigma-Aldrich

Diethylene glycol

SAJ first grade, ≥98.0%

Synonym(s):

2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
Beilstein:
969209
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

442 °F

expl. lim.

2-12.3 %

availability

available only in Japan

refractive index

n20/D 1.447 (lit.)

pH

5.0-8 (20 °C, 500 g/L)

bp

245 °C (lit.)

mp

−10 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

SMILES string

OCCOCCO

InChI

1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

MTHSVFCYNBDYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Handa et al.
Plant & cell physiology, 56(8), 1490-1511 (2015-05-27)
Gene expression during arbuscular mycorrhizal development is highly orchestrated in both plants and arbuscular mycorrhizal fungi. To elucidate the gene expression profiles of the symbiotic association, we performed a digital gene expression analysis of Lotus japonicus and Rhizophagus irregularis using
Bichismita Sahu et al.
The Journal of organic chemistry, 76(14), 5614-5627 (2011-05-31)
Developed in the early 1990s, peptide nucleic acid (PNA) has emerged as a promising class of nucleic acid mimic because of its strong binding affinity and sequence selectivity toward DNA and RNA and resistance to enzymatic degradation by proteases and
Pavel Kucheryavy et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 710-716 (2012-12-20)
Magnetite nanoparticles in the size range of 3.2-7.5 nm were synthesized in high yields under variable reaction conditions using high-temperature hydrolysis of the precursor iron(II) and iron(III) alkoxides in diethylene glycol solution. The average sizes of the particles were adjusted
Jihoon Shin et al.
Biomacromolecules, 13(11), 3833-3840 (2012-10-16)
Renewable ABA triblock copolymers were prepared by sequential polymerization of the plant-based monomers menthide and α-methylene-γ-butyrolactone (MBL or tulipalin A). Ring-opening transesterification polymerization of menthide using diethylene glycol as an initiator gave α,ω-dihydroxy poly(menthide) (HO-PM-OH), which was converted to α,ω-dibromo
Luc Faucher et al.
Nanotechnology, 22(29), 295103-295103 (2011-06-23)
Ultra-small gadolinium oxide nanoparticles (US-Gd(2)O(3)) are used to provide 'positive' contrast effects in magnetic resonance imaging (MRI), and are being considered for molecular and cellular imaging applications. However, these nanoparticles can aggregate over time in aqueous medium, as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service