Skip to Content
Merck
All Photos(1)

Key Documents

05-1700

Sigma-Aldrich

Carborundum

CP

Synonym(s):

Silicon carbide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiC
CAS Number:
Molecular Weight:
40.10
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

reaction suitability

reagent type: catalyst
core: silicon

availability

available only in Japan

mp

2700 °C (lit.)

density

3.22 g/mL at 25 °C (lit.)

SMILES string

[C-]#[Si+]

InChI

1S/CSi/c1-2

InChI key

HBMJWWWQQXIZIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Vermeersch et al.
Cardiovascular revascularization medicine : including molecular interventions, 13(6), 316-320 (2012-11-21)
The aim of this MULTIBENE study was to evaluate the safety and efficacy of the silicon carbide coated cobalt chromium PRO-Kinetik coronary stent system in patients with single de novo coronary lesions. This prospective international multicenter study included 202 patients
Martial Hamon et al.
EuroIntervention : journal of EuroPCR in collaboration with the Working Group on Interventional Cardiology of the European Society of Cardiology, 8(9), 1006-1011 (2013-01-24)
To report the four-month and nine-month angiographic results as well as one-year clinical follow-up from the first-in-man study with the silicon carbide and sirolimus-eluting bioabsorbable polymer (poly-L-lactic acid (PLLA) polymer) -coated cobalt-chromium Orsiro stent. A group of 30 patients with
Bálint Somogyi et al.
Nanoscale, 4(24), 7720-7726 (2012-11-09)
Molecule-sized fluorescent emitters are much sought-after to probe biomolecules in living cells. We demonstrate here by time-dependent density functional calculations that the experimentally achievable 1-2 nm sized silicon carbide nanocrystals can emit light in the near-infrared region after introducing appropriate
Mónica Fernández-Perea et al.
Optics express, 20(21), 24018-24029 (2012-11-29)
We have developed new, Mg/SiC multilayer coatings with corrosion barriers which can be used to efficiently and simultaneously reflect extreme ultraviolet (EUV) radiation in single or multiple narrow bands centered at wavelengths in the spectral region from 25 to 80
A Oliveros et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 1643-1646 (2013-02-01)
Silicon Carbide (SiC), has been shown to be a bio- and hema-compatible substrate that could potentially be used in biosensor applications. The development of a viable biorecognition interface using SiC as the substrate material for bio-detection is described. Surface modification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service