Skip to Content
Merck

Skip To

03-0240

Barium chloride dihydrate

JIS special grade, ≥99.0%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
UNSPSC Code:
12352302
PubChem Substance ID:
EC Number:
233-788-1
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

JIS special grade

assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

5.0-8.0 (25 °C, 50 g/L)

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L



Still not finding the right product?

Explore all of our products under Barium chloride dihydrate


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Laura A Merriam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4614-4622 (2013-03-08)
After G-protein-coupled receptor activation and signaling at the plasma membrane, the receptor complex is often rapidly internalized via endocytic vesicles for trafficking into various intracellular compartments and pathways. The formation of signaling endosomes is recognized as a mechanism that produces
Fan Hu et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(12), 1051-1058 (2012-08-15)
Danshen (Salviae Miltiorrhizae Radix) and Gegen (Puerariae Lobatae Radix) have been widely used in treating cardiovascular diseases for thousands of years in China. The present study was carried out to evaluate the effects of a Danshen and Gegen decoction (DG)
Mirna González-González et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 893-894, 182-186 (2012-03-27)
Recovery and purification of stem cells are determining steps in order to obtain the purity and viability required for transplantation. In this context, immunochemical techniques have been widely preferred due to their high selectivity. CD133, a glycoprotein expressed by stem



Questions

Reviews

No rating value

Active Filters