Skip to Content

LRNTKB

Roche

KAPA LongRange

suitable for PCR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.55
UNSPSC Code:
41106300
Technique(s):
PCR: suitable
Usage:
sufficient for 100 reactions, sufficient for 250 reactions
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 reactions, sufficient for 250 reactions

shelf life

≤18 mo.

feature

Long Range PCR, dNTPs included, hotstart: no

packaging

pkg of 100 U (KK3006), pkg of 250 U (KK3005)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

General description

The KAPA Long Range PCR system is a blend of Taq DNA polymerase and an engineered archaeal (B-family) DNA polymerase possessing proofreading capability. This two-enzyme system is designed specifically to support long-range and/or sensitive PCR, and possess 3 – 4X higher fidelity than Taq polymerase. In the hot-start formulation, KAPA Long Range is combined with a proprietary antibody that inactivates the enzyme until the first denaturation step, eliminating spurious amplification products and increasing reaction efficiency and sensitivity.

KAPA Long Range is also supplied in 2X ReadyMix format with loading dye that contains all the components required for PCR except primers and template. The loading dye in the reaction buffer allows you to load PCR products directly onto the agarose gel with no extra steps.

Application

KAPA LongRange is suitable for:
  • Polymerase chain reaction (PCR) amplification of long targets and/or low amounts
  • Standard short- and mid-range PCR
  • T-A cloning
  • Targeted sample enrichment for next-generation DNA sequencing

Biochem/physiol Actions

Both KAPA Taq DNA polymerase and a modified archaeal (Type B) DNA polymerase possess 5′→3′ polymerase activity, but only KAPA Taq DNA polymerase possesses 5′→3′ exonuclease activity, and only the Type B DNA polymerase possesses 3′→5′ exonuclease (proofreading) activity. Reaction products generated with KAPA LongRange are polyA tailed and can be cloned into TA-cloning vectors.

Features and Benefits

Amplify long targets with high sensitivity:
  • Amplification of targets with high sensitivity and specificity up to 15 kb
  • High yields with low input DNA

Outperforms with complex targets:
  • Exhibits high yield, greater sensitivity and specificity where competitor long-range systems fail

Quick Notes:
  • KAPA LongRange HotStart is a blend of KAPA Taq HotStart and a proofreading DNA polymerase.
  • Suitable for short-, medium- and long-range PCR, and PCR with low amounts of template DNA.
  • Fidelity is 2–4 times better than Taq.
  • For short amplicons, replace Taq DNA polymerase for improvements in yield and/or fidelity.Amplify mid- to long-range targets with high yield and sensitivity.
  • KAPA LongRange Buffer (Mg-free) is designed for long-range, sensitive PCR. MgCl2 is supplied separately for optimization.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for longterm storage.

Analysis Note

Each batch of KAPA LongRange HotStart DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). The KAPA LongRange HotStart PCR system is subjected to stringent quality control tests, is free of contaminating exo- and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.


Still not finding the right product?

Explore all of our products under KAPA LongRange


Kit Components Only

Product No.
Description

  • KAPA LongRange DNA Polymerase 5 U/μL

  • KAPA LongRange Buffer (5X)

  • MgCl2 25 mM

  • KAPA dNTP Mix 10 mM each

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library