Key Documents
ZTLP0EDI1
Milli-Q
Elix® Electrodeionization (EDI) Module
Water Purification by Electrodeionization (EDI)
Select a Size
1 KIT
Inquire
This item is not able to be purchased as a guest user. For further information, please contact us at +420246003252.
Select a Size
Change View
1 KIT
Inquire
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
This item is not able to be purchased as a guest user. For further information, please contact us at +420246003252.
packaging
pkg of 1 unit
manufacturer/tradename
Elix®
unit H
33 cm (13 in.)
General description
The Elix® EDI module is designed to continuously deionize the water, post-reverse osmosis by electrodeionization technique. The Elix® EDI module delivers superior quality pure water consistently and reliably, without any maintenance.
Application
The Type 2 water produced post RO-EDI treatment meets the needs of many general laboratory applications, and for use in clinical analyzers, cell culture incubators and weatherometers.
Features and Benefits
  • Efficiently removes ions.
  • High water recovery.
  • Low maintenance.
Legal Information
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany